[1-(3-Thienyl)cyclopropyl]amine hydrochloride

Supplier: Matrix Scientific

060922-500MG
101920-948EA 969.23 USD
101920-948
[1-(3-Thienyl)cyclopropyl]amine hydrochloride
[1-(3-Thienyl)cyclopropyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNS
MDL Number: MFCD19103327
CAS Number: 1332531-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR