[1-(2-Thienyl)cyclopropyl]amine hydrochloride

Supplier: Matrix Scientific

060921-500MG
101920-946EA 546.15 USD
101920-946
[1-(2-Thienyl)cyclopropyl]amine hydrochloride
[1-(2-Thienyl)cyclopropyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNS
MDL Number: MFCD19103326
CAS Number: 1332530-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR