[1-(2-Furyl)cyclopropyl]amine hydrochloride

Supplier: Matrix Scientific

060920-500MG
101920-944EA 449.23 USD
101920-944
[1-(2-Furyl)cyclopropyl]amine hydrochloride
[1-(2-Furyl)cyclopropyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNO
MDL Number: MFCD19103325
CAS Number: 1332529-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR