[2-(1,2-Oxazinan-2-yl)ethyl]amine dihydrochloride dihydrate

Supplier: Matrix Scientific

060917-500MG
101920-938EA 484.62 USD
101920-938
[2-(1,2-Oxazinan-2-yl)ethyl]amine dihydrochloride dihydrate
[2-(1,2-Oxazinan-2-yl)ethyl]amine dihydrochloride dihydrate

Hazard: Irritant

Formula: C₆H₂₀Cl₂N₂O₃
MDL Number: MFCD19103322
CAS Number: 1452488-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR