Methyl (3-amino-1H-1,2,4-triazol-5-yl)acetate hydrochloride

Supplier: Matrix Scientific

060913-500MG
101920-930EA 606.15 USD
101920-930
Methyl (3-amino-1H-1,2,4-triazol-5-yl)acetate hydrochloride
Methyl (3-amino-1H-1,2,4-triazol-5-yl)acetate hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₄O₂
MDL Number: MFCD19103320
CAS Number: 1279219-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR