(S)-Methyl-2-aminopentanoate hydrochloride
Supplier: Matrix Scientific
Synonyms:
L-Norvaline methyl ester hydrochloride
, H-Nva-OMe·HCl
060907-500MG
101920-920EA
243.08
USD
101920-920
(S)-Methyl-2-aminopentanoate hydrochloride
(S)-Methyl-2-aminopentanoate hydrochloride
Hazard: Irritant
Formula:
C₆H₁₄ClNO₂ MW: 167.633 g/mol |
MDL Number:
MFCD00190974 CAS Number: 56558-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...