4-Methoxy-2-piperazin-1-ylpyrimidine dihydrochloride

Supplier: Matrix Scientific

060904-500MG
101920-914EA 364.62 USD
101920-914
4-Methoxy-2-piperazin-1-ylpyrimidine dihydrochloride
4-Methoxy-2-piperazin-1-ylpyrimidine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₆Cl₂N₄O
MDL Number: MFCD12197370
CAS Number: 55745-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR