2-Chloro-5-methyl-1H-benzimidazole hydrochloride

Supplier: Matrix Scientific

060901-500MG
101920-908EA 438.46 USD
101920-908
2-Chloro-5-methyl-1H-benzimidazole hydrochloride
2-Chloro-5-methyl-1H-benzimidazole hydrochloride

Hazard: Irritant

Formula: C₈H₈Cl₂N₂
MW: 203.07 g/mol
MDL Number: MFCD19104827
CAS Number: 4887-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR