1-(1-Adamantyl)-5-methyl-1,2,3,6-tetrahydropyridine hydrochloride

Supplier: Matrix Scientific

060883-500MG
101920-872EA 364.62 USD
101920-872
1-(1-Adamantyl)-5-methyl-1,2,3,6-tetrahydropyridine hydrochloride
1-(1-Adamantyl)-5-methyl-1,2,3,6-tetrahydropyridine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClN
MDL Number: MFCD19103296
CAS Number: 1332531-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR