3-(1-Adamantyloxy)pyridine hydrobromide

Supplier: Matrix Scientific

060881-500MG
101920-868EA 243.08 USD
101920-868
3-(1-Adamantyloxy)pyridine hydrobromide
3-(1-Adamantyloxy)pyridine hydrobromide

Hazard: Irritant

Formula: C₁₅H₂₀BrNO
MDL Number: MFCD19103294
CAS Number: 1332528-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR