1-(1-Adamantyl)-3-methylpyridinium bromide

Supplier: Matrix Scientific

060876-500MG
101920-858EA 243.08 USD
101920-858
1-(1-Adamantyl)-3-methylpyridinium bromide
1-(1-Adamantyl)-3-methylpyridinium bromide

Hazard: Irritant

Formula: C₁₆H₂₂BrN
MDL Number: MFCD19103290
CAS Number: 112671-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR