(2-Hydroxy-4,5-dimethylphenyl)-N,N,N-trimethylmethanaminium iodide

Supplier: Matrix Scientific

060873-500MG
101920-852EA 402.5 USD
101920-852
(2-Hydroxy-4,5-dimethylphenyl)-N,N,N-trimethylmethanaminium iodide
(2-Hydroxy-4,5-dimethylphenyl)-N,N,N-trimethylmethanaminium iodide

Hazard: Irritant

Formula: C₁₂H₂₀INO
MDL Number: MFCD19103283
CAS Number: 1210475-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR