(4-Methoxy-2-methylbenzyl)amine hydrochloride

Supplier: Matrix Scientific

060870-500MG
101920-846EA 364.62 USD
101920-846
(4-Methoxy-2-methylbenzyl)amine hydrochloride
(4-Methoxy-2-methylbenzyl)amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClNO
MDL Number: MFCD11045201
CAS Number: 856627-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR