1-Pyrazin-2-ylpiperidin-3-amine dihydrochloride

Supplier: Matrix Scientific

060869-500MG
101920-844EA 484.62 USD
101920-844
1-Pyrazin-2-ylpiperidin-3-amine dihydrochloride
1-Pyrazin-2-ylpiperidin-3-amine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₆Cl₂N₄
MDL Number: MFCD19103266
CAS Number: 1185319-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR