Ethyl-1-aminocyclopentanecarboxylate hydrochloride

Supplier: Matrix Scientific

060862-500MG
101920-830EA 606.15 USD
101920-830
Ethyl-1-aminocyclopentanecarboxylate hydrochloride
Ethyl-1-aminocyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD01735325
CAS Number: 22649-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR