3-Piperidin-4-ylpropanoic acid hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-(Piperidin-4-yl)propanoic acid hydrochloride

060861-500MG
101920-828EA 484.62 USD
101920-828
3-Piperidin-4-ylpropanoic acid hydrochloride
3-Piperidin-4-ylpropanoic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MW: 193.68 g/mol
MDL Number: MFCD06202371
CAS Number: 1822-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR