2-(4-Methylphenyl)-4,5,6,7-tetrahydro[1,3]thiazolo[5,4-c]pyridine hydrobromide

Supplier: Matrix Scientific

060857-500MG
101920-820EA 484.62 USD
101920-820
2-(4-Methylphenyl)-4,5,6,7-tetrahydro[1,3]thiazolo[5,4-c]pyridine hydrobromide
2-(4-Methylphenyl)-4,5,6,7-tetrahydro[1,3]thiazolo[5,4-c]pyridine hydrobromide

Hazard: Irritant

Formula: C₁₃H₁₆Br₂N₂S
MDL Number: MFCD18071281
CAS Number: 1332530-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR