5-(Aminomethyl)-2-phenyl-1,2-dihydro-3H-pyrazol-3-one dihydrochloride

Supplier: Matrix Scientific

060856-500MG
101920-818EA 606.15 USD
101920-818
5-(Aminomethyl)-2-phenyl-1,2-dihydro-3H-pyrazol-3-one dihydrochloride
5-(Aminomethyl)-2-phenyl-1,2-dihydro-3H-pyrazol-3-one dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃Cl₂N₃O
MDL Number: MFCD09790129
CAS Number: 1052552-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR