2-Phenylmorpholine hydrochloride

Supplier: Matrix Scientific

Synonyms: (Morpholin-2-yl)benzene hydrochloride

060855-500MG
101920-816EA 283.08 USD
101920-816
2-Phenylmorpholine hydrochloride
2-Phenylmorpholine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃NO·ClH
MW: 199.677 g/mol
MDL Number: MFCD03869024
CAS Number: 23972-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR