N,N-Dimethyl-N'-[2-(trifluoromethoxy)benzyl]ethane-1,2-diamine dihydrochloride

Supplier: Matrix Scientific

060851-500MG
101920-808EA 364.62 USD
101920-808
N,N-Dimethyl-N'-[2-(trifluoromethoxy)benzyl]ethane-1,2-diamine dihydrochloride
N,N-Dimethyl-N'-[2-(trifluoromethoxy)benzyl]ethane-1,2-diamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉Cl₂F₃N₂O
MW: 335.2 g/mol
MDL Number: MFCD18071375
CAS Number: 1328641-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR