N'-(1-Benzylpiperidin-4-yl)-N,N-dimethylethane-1,2-diamine dihydrochloride

Supplier: Matrix Scientific

060849-500MG
101920-804EA 364.62 USD
101920-804
N'-(1-Benzylpiperidin-4-yl)-N,N-dimethylethane-1,2-diamine dihydrochloride
N'-(1-Benzylpiperidin-4-yl)-N,N-dimethylethane-1,2-diamine dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₉Cl₂N₃
MDL Number: MFCD18071374
CAS Number: 1332531-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR