N-(Dibenzo[b,d]furan-4-ylmethyl)propan-2-amine hydrochloride

Supplier: Matrix Scientific

060845-500MG
101920-796EA 606.15 USD
101920-796
N-(Dibenzo[b,d]furan-4-ylmethyl)propan-2-amine hydrochloride
N-(Dibenzo[b,d]furan-4-ylmethyl)propan-2-amine hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈ClNO
MDL Number: MFCD09759082
CAS Number: 1189685-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR