5-(Chloromethyl)-2-(4-methoxyphenyl)-1,3-thiazole hydrochloride

Supplier: Matrix Scientific

060843-500MG
101920-792EA 727.69 USD
101920-792
5-(Chloromethyl)-2-(4-methoxyphenyl)-1,3-thiazole hydrochloride
5-(Chloromethyl)-2-(4-methoxyphenyl)-1,3-thiazole hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₁Cl₂NOS
MDL Number: MFCD18071361
CAS Number: 1332528-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR