N-[(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methyl]cyclopropanamine dihydrochloride

Supplier: Matrix Scientific

060842-500MG
101920-790EA 727.69 USD
101920-790
N-[(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methyl]cyclopropanamine dihydrochloride
N-[(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methyl]cyclopropanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈Cl₂N₂S
MW: 317.28 g/mol
MDL Number: MFCD18071355
CAS Number: 1332529-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR