(2-Chloro-4,5-dimethoxybenzyl)amine

Supplier: Matrix Scientific

058432-500MG
101914-798EA 306.15 USD
101914-798
(2-Chloro-4,5-dimethoxybenzyl)amine
(2-Chloro-4,5-dimethoxybenzyl)amine

Hazard: Irritant

Formula: C₉H₁₂ClNO₂
MDL Number: MFCD17676107
CAS Number: 1246471-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR