DIPSO

Supplier: Matrix Scientific

Synonyms: 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulfonic acid3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid

058427-500MG
101914-790EA 306.15 USD
101914-790
DIPSO
DIPSO

Hazard: Irritant

Formula: C₇H₁₇NO₆S
MW: 261.29 g/mol
MDL Number: MFCD00038353
CAS Number: 68399-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR