1-(Cyclopropylcarbonyl)-4-piperidinecarboxamide

Supplier: Matrix Scientific

058415-500MG
101914-764EA 263.08 USD
101914-764
1-(Cyclopropylcarbonyl)-4-piperidinecarboxamide
1-(Cyclopropylcarbonyl)-4-piperidinecarboxamide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂
MDL Number: MFCD11643143
CAS Number: 1092023-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR