N-(2-Furylmethyl)-3-methyl-2-(1H-pyrrol-1-yl)butanamide

Supplier: Matrix Scientific

058414-500MG
101914-762EA 306.15 USD
101914-762
N-(2-Furylmethyl)-3-methyl-2-(1H-pyrrol-1-yl)butanamide
N-(2-Furylmethyl)-3-methyl-2-(1H-pyrrol-1-yl)butanamide

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₂
MW: 246.31 g/mol
MDL Number: MFCD08083045
CAS Number: 887029-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR