N-(3,4-Dimethylphenyl)-3-methyl-2-(1H-pyrrol-1-yl)butanamide

Supplier: Matrix Scientific

058413-500MG
101914-760EA 306.15 USD
101914-760
N-(3,4-Dimethylphenyl)-3-methyl-2-(1H-pyrrol-1-yl)butanamide
N-(3,4-Dimethylphenyl)-3-methyl-2-(1H-pyrrol-1-yl)butanamide

Hazard: Irritant

Formula: C₁₇H₂₂N₂O
MDL Number: MFCD08083044
CAS Number: 887029-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR