5-(2-Furyl)-1H-pyrazole-3-carboxamide
Supplier: Matrix Scientific
058408-500MG
101914-750EA
115.38
USD
101914-750
5-(2-Furyl)-1H-pyrazole-3-carboxamide
5-(2-Furyl)-1H-pyrazole-3-carboxamide
Hazard: Irritant
Formula:
C₈H₇N₃O₂ |
MDL Number:
MFCD13250139 CAS Number: 857283-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...