2-(3-Methylbutoxy)benzoic acid

Supplier: Matrix Scientific

058398-500MG
101914-730EA 306.15 USD
101914-730
2-(3-Methylbutoxy)benzoic acid
2-(3-Methylbutoxy)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₃
MDL Number: MFCD06800853
CAS Number: 21023-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR