(4-Chlorophenyl)(2-furyl)methanone

Supplier: Matrix Scientific

058395-500MG
101914-722EA 129.23 USD
101914-722
(4-Chlorophenyl)(2-furyl)methanone
(4-Chlorophenyl)(2-furyl)methanone

Hazard: Irritant

Formula: C₁₁H₇ClO₂
MDL Number: MFCD03144309
CAS Number: 13365-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR