2-[1-(3-Methylbenzyl)-2-piperazinyl]ethanol

Supplier: Matrix Scientific

058394-500MG
101914-720EA 306.15 USD
101914-720
2-[1-(3-Methylbenzyl)-2-piperazinyl]ethanol
2-[1-(3-Methylbenzyl)-2-piperazinyl]ethanol

Hazard: Irritant

Formula: C₁₄H₂₂N₂O
MDL Number: MFCD05870522
CAS Number: 815655-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR