2-(1-Benzyl-2-piperazinyl)ethanol dihydrochloride

Supplier: Matrix Scientific

058392-500MG
101914-716EA 306.15 USD
101914-716
2-(1-Benzyl-2-piperazinyl)ethanol dihydrochloride
2-(1-Benzyl-2-piperazinyl)ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₂Cl₂N₂O
MDL Number: MFCD17012619
CAS Number: 1233642-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR