2,4-Dichloro-5-nitrobenzoic acid

Supplier: Matrix Scientific

058380-500MG
101914-694EA 138.46 USD
101914-694
2,4-Dichloro-5-nitrobenzoic acid
2,4-Dichloro-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₃Cl₂NO₄
MDL Number: MFCD00456403
CAS Number: 19861-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR