4,4'-[(4-Fluorophenyl)methylene]dimorpholine

Supplier: Matrix Scientific

058361-500MG
101914-654EA 233.85 USD
101914-654
4,4'-[(4-Fluorophenyl)methylene]dimorpholine
4,4'-[(4-Fluorophenyl)methylene]dimorpholine

Hazard: Irritant

Formula: C₁₅H₂₁FN₂O₂
MDL Number: MFCD00424679
CAS Number: 6425-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR