4-(Aminomethyl)benzenesulfonamide acetate

Supplier: Matrix Scientific

058350-500MG
101914-634EA 306.15 USD
101914-634
4-(Aminomethyl)benzenesulfonamide acetate
4-(Aminomethyl)benzenesulfonamide acetate

Hazard: Irritant

Formula: C₉H₁₄N₂O₄S
MW: 246.2835 g/mol
MDL Number: MFCD00072089
CAS Number: 13009-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR