1(2H)-Phthalazinone

Supplier: Matrix Scientific

Synonyms: Phthalazin-1(2H)-one

058345-500MG
101914-624EA 306.15 USD
101914-624
1(2H)-Phthalazinone
1(2H)-Phthalazinone

Hazard: Irritant

Formula: C₈H₆N₂O
MW: 146.1500 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00006892
CAS Number: 119-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR