Methyl [4-(chlorosulfonyl)phenyl]carbamate

Supplier: Matrix Scientific

058344-500MG
101914-622EA 306.15 USD
101914-622
Methyl [4-(chlorosulfonyl)phenyl]carbamate
Methyl [4-(chlorosulfonyl)phenyl]carbamate

Hazard: Irritant

Formula: C₈H₈ClNO₄S
MDL Number: MFCD00486540
CAS Number: 21926-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR