Diethyl (acetyloxy)malonate

Supplier: Matrix Scientific

058339-500MG
101914-614EA 306.15 USD
101914-614
Diethyl (acetyloxy)malonate
Diethyl (acetyloxy)malonate

Hazard: Irritant

Formula: C₉H₁₄O₆
MDL Number: MFCD00039895
CAS Number: 5468-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR