1-Azabicyclo[2.2.2]oct-3-yl(diphenyl)methanol hydrochloride

Supplier: Matrix Scientific

058337-500MG
101914-610EA 236.92 USD
101914-610
1-Azabicyclo[2.2.2]oct-3-yl(diphenyl)methanol hydrochloride
1-Azabicyclo[2.2.2]oct-3-yl(diphenyl)methanol hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₄ClNO
MDL Number: MFCD00467913
CAS Number: 10447-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR