6-Methylpiperidine-2-carboxylic acid hydrochloride

Supplier: Matrix Scientific

058336-500MG
101914-608EA 306.15 USD
101914-608
6-Methylpiperidine-2-carboxylic acid hydrochloride
6-Methylpiperidine-2-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO₂
MW: 179.644 g/mol
MDL Number: MFCD16885718
CAS Number: 99571-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR