9-Methoxy-4-methyl-2,4,5,6-tetrahydro-1H-3,4,6a-triazafluoranthene hydrochloride

Supplier: Matrix Scientific

058329-500MG
101914-598EA 306.15 USD
101914-598
9-Methoxy-4-methyl-2,4,5,6-tetrahydro-1H-3,4,6a-triazafluoranthene hydrochloride
9-Methoxy-4-methyl-2,4,5,6-tetrahydro-1H-3,4,6a-triazafluoranthene hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₈ClN₃O
MDL Number: MFCD00420801
CAS Number: 53734-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR