6-Cyclohexyl-2,9-dihydro-1H-carbazol-1-one

Supplier: Matrix Scientific

058326-500MG
101914-592EA 306.15 USD
101914-592
6-Cyclohexyl-2,9-dihydro-1H-carbazol-1-one
6-Cyclohexyl-2,9-dihydro-1H-carbazol-1-one

Hazard: Irritant

Formula: C₁₈H₁₉NO
MDL Number: MFCD16885719
CAS Number: 1233641-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR