2,3-Dihydroxynaphthoquinone

Supplier: Matrix Scientific

058322-500MG
101914-586EA 306.15 USD
101914-586
2,3-Dihydroxynaphthoquinone
2,3-Dihydroxynaphthoquinone

Hazard: Irritant

Formula: C₁₀H₆O₄
Melting Pt: 282 °C
MDL Number: MFCD00019554
CAS Number: 605-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR