Methyl-2,3-dibromo-2-methylpropanoate

Supplier: Matrix Scientific

058313-500MG
101914-566EA 306.15 USD
101914-566
Methyl-2,3-dibromo-2-methylpropanoate
Methyl-2,3-dibromo-2-methylpropanoate

Hazard: Irritant

Formula: C₅H₈Br₂O₂
MDL Number: MFCD00154484
CAS Number: 3673-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR