4-Methoxy-2-methylaniline hydrochloride

Supplier: Matrix Scientific

058304-500MG
101914-548EA 306.15 USD
101914-548
4-Methoxy-2-methylaniline hydrochloride
4-Methoxy-2-methylaniline hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClNO
MW: 173.6400 g/mol
MDL Number: MFCD00155304
CAS Number: 133985-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR