Methyl benzylcarbamate

Supplier: Matrix Scientific

058279-500MG
101914-498EA 292.31 USD
101914-498
Methyl benzylcarbamate
Methyl benzylcarbamate

Hazard: Irritant

Formula: C₉H₁₁NO₂
MDL Number: MFCD00025856
CAS Number: 5817-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR