tert-Butyl-4-[(4-fluoro-5-methyl-2-nitrophenyl)amino]piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
058266-500MG
058266-1G
101914-460EA
467.69
USD
101914-460
101914-458
tert-Butyl-4-[(4-fluoro-5-methyl-2-nitrophenyl)amino]piperidine-1-carboxylate ≥95%
tert-Butyl-4-[(4-fluoro-5-methyl-2-nitrophenyl)amino]piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₇H₂₄FN₃O₄ MW: 353.4 g/mol |
MDL Number:
MFCD17167312 CAS Number: 932374-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...