1-Methyl-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

058251-500MG
101914-402EA 1096.92 USD
101914-402
1-Methyl-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylic acid ≥95%
1-Methyl-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₄
MDL Number: MFCD11203524
CAS Number: 1038385-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR